accessibility menu, dialog, popup

UserName

CAS RN 194017-70-4

IUPAC Name: [4-(pyridin-2-yloxy)phenyl]methanol
Synonyms: [4-(pyridin-2-yloxy)phenyl]methanol
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: ZDTXOQRCIVCMLT-UHFFFAOYSA-N
SMILES: OCC1=CC=C(OC2=CC=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results