accessibility menu, dialog, popup

UserName

CH3OHNClOHO

CAS RN 19407-42-2

IUPAC Name: 2-chloro-6-acetamidobenzoic acid
Synonyms: 2-chloro-6-acetamidobenzoic acid
Molecular Weight (g/mol): 213.62
Molecular Formula: C9H8ClNO3
InChi Key: VFHSJTHAMJFUCK-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(Cl)=C1C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results