accessibility menu, dialog, popup

UserName

ClHNSHNCl

CAS RN 19462-98-7

IUPAC Name: 5,6-dichloro-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 5,6-dichloro-1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 219.08
Molecular Formula: C7H4Cl2N2S
InChi Key: AFDOMGKBKBKUHB-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C=C2NC(=S)NC2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results