missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dichloro-2-mercaptobenzimidazole, 98%

Product Code. 11446847
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446847 5 g 5g
11436847 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446847 Supplier Thermo Scientific Alfa Aesar Supplier No. A14309.06
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19462-98-7 | C7H4Cl2N2S | 219.083 g/mol

It is employed in the synthesis of 2?,3?-dideoxy and 2?,3?-unsaturated ribofuranonucleosides as potential antiviral agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of 2′,3′-dideoxy and 2′,3′-unsaturated ribofuranonucleosides as potential antiviral agents.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19462-98-7
Molecular Formula C7H4Cl2N2S
Molecular Weight (g/mol) 219.083
MDL Number MFCD00052398
InChI Key AFDOMGKBKBKUHB-UHFFFAOYSA-N
PubChem CID 2774259
IUPAC Name 5,6-dichloro-1,3-dihydrobenzimidazole-2-thione
SMILES C1=C2C(=CC(=C1Cl)Cl)NC(=S)N2

Specifications

Melting Point ∼350°C (decomposition)
Odor Odorless
Quantity 5 g
Synonym 5,6-dichloro-1h-benzo d imidazole-2-thiol, 5,6-dichlorobenzimidazole-2-thiol, 5,7dicl2sh-bzind, 2-mercapto-5,6-dichlorobenzimidazole, 5,6-dichloro-1h-benzimidazole-2-thiol, 5,6-dichloro-2-mercaptobenzimidazole, 5,6-dichloro-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione,5,6-dichloro-1,3-dihydro, 2h-benzimidazole-2-thione, 5,6-dichloro-1,3-dihydro, 5,6-dichloro-1,3-dihydro-1,3-benzodiazole-2-thione
Solubility Information Insoluble in water.
Formula Weight 219.09
Percent Purity 98%
Chemical Name or Material 5,6-Dichloro-2-mercaptobenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.