accessibility menu, dialog, popup

UserName

CAS RN 19472-74-3

BrN

CAS RN 19472-74-3

IUPAC Name: 2-(2-bromophenyl)acetonitrile
Synonyms: (2-bromo-phenyl)-acetonitrile
Molecular Weight (g/mol): 196.05
Molecular Formula: C8H6BrN
InChi Key: BVCOJESIQPNOIF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results