accessibility menu, dialog, popup

UserName

FNFFF

CAS RN 194853-86-6

IUPAC Name: 4-fluoro-2-(trifluoromethyl)benzonitrile
Synonyms: 4-fluoro-2-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 189.11
Molecular Formula: C8H3F4N
InChi Key: LCCPQUYXMFXCAC-UHFFFAOYSA-N
SMILES: FC1=CC=C(C#N)C(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results