accessibility menu, dialog, popup

UserName

CAS RN 19524-06-2

IUPAC Name: 4-bromopyridine hydrochloridyl
Synonyms: 4-bromopyridine hydrochloridyl
Molecular Weight (g/mol): 193.45
Molecular Formula: C5H4BrClN
InChi Key: PSPIMDWZDKGZBO-UHFFFAOYSA-N
SMILES: [Cl].BrC1=CC=NC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)