missing translation for 'onlineSavingsMsg'
Learn More

4-Bromopyridine hydrochloride, 99%

Product Code. 11337116
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337116 25 g 25g
11317116 1 g 1g
11327116 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337116 Supplier Thermo Scientific Alfa Aesar Supplier No. L07052.14
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19524-06-2 | C5H5BrClN | 194.456 g/mol

4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in DMSO, Methanol and Water.

Notes
Store at -20°C. Hygroscopic. Incompatible materials are moisture, strong bases, strong oxidizing agents. Protect from humidity and air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19524-06-2
Molecular Formula C5H5BrClN
Molecular Weight (g/mol) 194.456
MDL Number MFCD00012828
InChI Key MPZMVUQGXAOJIK-UHFFFAOYSA-N
Synonym 4-bromopyridine hydrochloride, 4-bromopyridinium chloride, 4-bromopyridine hcl, pyridine, 4-bromo-, hydrochloride, 4-bromo-pyridine hcl, 4-bromo pyridine hcl, 4-bromopyridine, chloride, 4-bromopyridinehydrochloride, 4-bromopyridine hydrochloride salt, 4-bromopyridine-hcl
PubChem CID 88100
IUPAC Name 4-bromopyridine;hydrochloride
SMILES C1=CN=CC=C1Br.Cl

Specifications

Melting Point ∽270°C (decomposition)
Quantity 25 g
Beilstein 3621847
Sensitivity Hygroscopic
Solubility Information Soluble in DMSO,Methanol and Water.
Formula Weight 194.46
Percent Purity 99%
Chemical Name or Material 4-Bromopyridine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.