accessibility menu, dialog, popup

UserName

CAS RN 195447-79-1

FFFFOHO

CAS RN 195447-79-1

IUPAC Name: 2-[3-fluoro-5-(trifluoromethyl)phenyl]acetic acid
Synonyms: [3-fluoro-5-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: LQIBHDUPSIQRPV-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(=CC(F)=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results