accessibility menu, dialog, popup

UserName

H2CO

CAS RN 19600-63-6

IUPAC Name: 2-(hex-5-en-1-yl)oxirane
Synonyms: 2-(hex-5-en-1-yl)oxirane
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: UKTHULMXFLCNAV-UHFFFAOYNA-N
SMILES: C=CCCCCC1CO1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)