missing translation for 'onlineSavingsMsg'
Learn More

1,2-Epoxy-7-octene, 97%, Thermo Scientific™

Product Code. 10682942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10682942 25 g Glass bottle 25g
10643472 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10682942 Supplier Thermo Scientific Acros Supplier No. 335140250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 19600-63-6
MDL Number MFCD00005156
InChI Key UKTHULMXFLCNAV-UHFFFAOYSA-N
Synonym 1,2-epoxy-7-octene, oxirane, 5-hexenyl, 5-hexenyloxirane, 7,8-epoxyoctene, ccris 3749, 2-hex-5-en-1-yl oxirane, oxirane, 5-hexenyl-, s, 2-hex-5-en-yloxirane, 2-5-hexenyl oxirane, acmc-20ap64
PubChem CID 29678
IUPAC Name 2-hex-5-enyloxirane
SMILES C=CCCCCC1CO1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8500g/mL
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H14O
Refractive Index 1.4345 to 1.4365
Quantity 25 g
Specific Gravity 0.85
Solubility Information Solubility in water: immiscible.
Molecular Weight (g/mol) 126.199
Formula Weight 126.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 2-Epoxy-7-octene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,2-Epoxy-7-octene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.