accessibility menu, dialog, popup

UserName

CH3NOOHO

CAS RN 19668-85-0

IUPAC Name: 2-(3-methyl-1,2-oxazol-5-yl)acetic acid
Synonyms: (3-methyl-1,2-oxazol-5-yl)acetic acid
Molecular Weight (g/mol): 141.13
Molecular Formula: C6H7NO3
InChi Key: POEFJFLAFQWOTL-UHFFFAOYSA-N
SMILES: CC1=NOC(CC(O)=O)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results