missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-isoxazoleacetic acid, 98%, Thermo Scientific™

Product Code. 10664375 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10664375 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10664375 Supplier Thermo Scientific Acros Supplier No. 426650010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19668-85-0
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.13
MDL Number MFCD01863539
InChI Key POEFJFLAFQWOTL-UHFFFAOYSA-N
Synonym 2-3-methylisoxazol-5-yl acetic acid, 3-methyl-5-isoxazoleacetic acid, 5-isoxazoleacetic acid, 3-methyl, 2-3-methyl-1,2-oxazol-5-yl acetic acid, 3-methyl-1,2-oxazol-5-yl acetic acid, 5-isoxazoleacetic acid,3-methyl, zlchem 1304, 3-methyl-5-isoxazole acetic acid, 3-methyl-isoxazol-5-yl-acetic acid, 5-carboxymethyl-3-methylisoxazole
PubChem CID 4195532
IUPAC Name 2-(3-methyl-1,2-oxazol-5-yl)acetic acid
SMILES CC1=NOC(=C1)CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 101.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 141.13
Percent Purity 98%
Physical Form Needle-like Crystals or Fine Crystalline Powder
Chemical Name or Material 3-Methyl-5-isoxazoleacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-5-isoxazoleacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.