CAS RN 19781-29-4
CAS RN 19781-29-4
IUPAC Name:
3-ethyloctan-4-one
Synonyms:
3-ethyloctan-4-one
Molecular Weight (g/mol):
156.27
Molecular Formula:
C10H20O
InChi Key:
XIQNUWMKJWNUCQ-UHFFFAOYSA-N
SMILES:
CCCCC(=O)C(CC)CC
1
–
1
of
1
results
1
3-Ethyl-4-octanone, 98%, Thermo Scientific™
CAS: 19781-29-4 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.269 MDL Number: MFCD00048806 InChI Key: XIQNUWMKJWNUCQ-UHFFFAOYSA-N Synonym: 3-ethyl-4-octanone,acmc-20an4b,4-octanone, 3-ethyl PubChem CID: 140602 IUPAC Name: 3-ethyloctan-4-one SMILES: CCCCC(=O)C(CC)CC