accessibility menu, dialog, popup

UserName

ClNH2(S)(R)H

CAS RN 1986-47-6

IUPAC Name: hydrogen (1S,2R)-2-phenylcyclopropan-1-amine chloride
Synonyms: hydrogen (1S,2R)-2-phenylcyclopropan-1-amine chloride
Molecular Weight (g/mol): 169.65
Molecular Formula: C9H12ClN
InChi Key: ZPEFMSTTZXJOTM-RJUBDTSPSA-N
SMILES: [H+].[Cl-].N[C@H]1C[C@@H]1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)