missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Tranylcypromine hydrochloride
Description
Irreversible inhibitor of lysine-specific demethylase 1 (LSD1/BHC110) and monoamine oxidase (MAO). Inhibits histone demethylation. In combination with CHIR 99021 (Cat. No. 4423), enables reprogramming of mouse embryonic fibroblasts transduced by only two factors, Oct4 and Klf4, into induced pluripotent stem (iPS) cells.
Specifications
Specifications
| CAS | 1986-47-6 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Molecular Formula | C9H12ClN |
| Formula Weight | 169.65 |
| Purity | >98% |
| Molecular Weight (g/mol) | 169.652 |
| Chemical Name or Material | Tranylcypromine hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?