accessibility menu, dialog, popup

UserName

CAS RN 1992-15-0

FFFFFFOHFFFFFFOH

CAS RN 1992-15-0

IUPAC Name: 1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
Synonyms: 1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
Molecular Weight (g/mol): 410.16
Molecular Formula: C12H6F12O2
InChi Key: YTJDSANDEZLYOU-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)