missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene, 98%, Thermo Scientific Chemicals

Product Code. 11439746
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439746 10 g 10g
11349246 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439746 Supplier Thermo Scientific Alfa Aesar Supplier No. B20259.09
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1992-15-0 | Molecular Formula: C12H6F12O2 | Molecular Weight: 410.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1992-15-0
Molecular Formula C12H6F12O2
Molecular Weight (g/mol) 410.16
MDL Number MFCD00042091
InChI Key YTJDSANDEZLYOU-UHFFFAOYSA-N
Synonym 1,4-bis hexafluoro-alpha-hydroxyisopropyl benzene, 2,2'-1,4-phenylene bis 1,1,1,3,3,3-hexafluoropropan-2-ol, 1,4-bis 2-hydroxyhexafluoroisopropyl benzene, 1,4-bis perfluoro 2-hydroxyprop-2-yl benzene, 1,1,1,3,3,3-hexafluoro-2-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl propan-2-ol, 1,4-bis 1,1,1,3,3,3-hexafluoro-2-hydroxypropyl benzene, alpha,alpha,alpha',alpha'-tetrakis trifluoromethyl-1,4-benzenedimethanol, 1,1,1,3,3,3-hexafluoro-2-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethy l phenyl propan-2-ol, 2,2'-p-phenylene bis 1,1,1,3,3,3-hexafluoro-2-propanol, 1,4-bis 2-hydroxyhexafluoro-2-propyl benzene
PubChem CID 631065
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]propan-2-ol
SMILES OC(C1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 92°C to 95°C
Boiling Point 100°C (20 mmHg)
Quantity 10 g
Formula Weight 410.16
Percent Purity 98%
Chemical Name or Material 1,4-Bis(2-hydroxyhexafluoroisopropyl)benzene

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.