accessibility menu, dialog, popup

UserName

CAS RN 19947-75-2

CH3NOH2NH3C

CAS RN 19947-75-2

IUPAC Name: dimethyl-1,2-oxazol-5-amine
Synonyms: dimethyl-1,2-oxazol-5-amine
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: PYNDWPFZDQONDV-UHFFFAOYSA-N
SMILES: CC1=NOC(N)=C1C
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)