missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylisoxazol-5-amine, 97%, Thermo Scientific™

Product Code. 10703643
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10703643 1 g 1g
10120045 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10703643 Supplier Thermo Scientific Maybridge Supplier No. SB00914DA
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 19947-75-2
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD00046081
InChI Key PYNDWPFZDQONDV-UHFFFAOYSA-N
Synonym 5-amino-3,4-dimethylisoxazole, 3,4-dimethylisoxazol-5-amine, 5-amino-3,4-dimethyl-isoxazole, 5-isoxazolamine, 3,4-dimethyl, dimethyl-1,2-oxazol-5-amine, 3,4-dimethyl-5-isoxazolamine, 5-amino-3,4-dimethyl isoxazole, 3,4-dimethylisoxazole-5-ylamine, 3,4-dimethylisoxazol-5-ylamine, pubchem8660
PubChem CID 88317
IUPAC Name 3,4-dimethyl-1,2-oxazol-5-amine
SMILES CC1=C(ON=C1C)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 112.13
Percent Purity 97%
Chemical Name or Material 3, 4-Dimethylisoxazol-5-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.