CAS RN 199664-70-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-(2-Fluorophenyl)-5-oxovaleric Acid, TRC
CAS: 199664-70-5 Molecular Formula: C11H11FO3 Molecular Weight (g/mol): 210.2 Synonym: 2-Fluoro-delta-oxobenzenepentanoic Acid SMILES: OC(=O)CCCC(=O)c1ccccc1F