accessibility menu, dialog, popup

UserName

CAS RN 199678-12-1

NNOOH

CAS RN 199678-12-1

IUPAC Name: 4-(pyrimidin-2-yl)benzoic acid
Synonyms: 4-(pyrimidin-2-yl)benzoic acid
Molecular Weight (g/mol): 200.2
Molecular Formula: C11H8N2O2
InChi Key: WNDAEOTYLPWXPN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results