CAS RN 1998079-15-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(3-morpholino-5,6-dihydropyridine)diazene Apixaban, TRC
CAS: 1998079-15-4 Molecular Formula: C30 H34 N6 O4 Molecular Weight (g/mol): 542.629 Synonym: Apixaban Impurity 16,5-Acetyl-1-(4-methoxyphenyl)-4-(2-((4-(2-oxopiperidin-1-yl)phenyl)amino)ethyl)-1H-pyrazole-3-carboxamide IUPAC Name: 5-morpholin-4-yl-1-[4-[(E)-[4-(5-morpholin-4-yl-6-oxo-2,3-dihydropyridin-1-yl)phenyl]diazenyl]phenyl]-2,3-dihydropyridin-6-one SMILES: O=C1N(CCC=C1N2CCOCC2)c3ccc(cc3)\N=N\c4ccc(cc4)N5CCC=C(N6CCOCC6)C5=O