accessibility menu, dialog, popup

UserName

CAS RN 2004-03-7

CH3NNNHN

CAS RN 2004-03-7

IUPAC Name: 6-methyl-7H-purine
Synonyms: 6-methylpurine
Molecular Weight (g/mol): 134.14
Molecular Formula: C6H6N4
InChi Key: SYMHUEFSSMBHJA-UHFFFAOYSA-N
SMILES: CC1=C2NC=NC2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results