missing translation for 'onlineSavingsMsg'
Learn More

6-Methylpurine, TRC

Product Code. 30468252
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468252 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468252 Supplier Toronto Research Chemicals Supplier No. TRCM32595050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2004-03-7
Molecular Formula C6 H6 N4
Quantity 50 mg
InChI Formula InChI=1S/C6H6N4/c1-4-5-6(9-2-7-4)10-3-8-5/h2-3H,1H3,(H,7,8,9,10)
SMILES Cc1ncnc2[nH]cnc12
IUPAC Name 6-methyl-9H-purine
Molecular Weight (g/mol) 134.14
Formula Weight 134.0592
Chemical Name or Material 6-Methylpurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.