accessibility menu, dialog, popup

UserName

OH(R)O(R)(R)OH(S)OHNNClNN

CAS RN 2004-06-0

IUPAC Name: (2R,3R,4S,5R)-2-(6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: (2R,3R,4S,5R)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 286.67
Molecular Formula: C10H11ClN4O4
InChi Key: XHRJGHCQQPETRH-KQYNXXCUSA-N
SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(Cl)N=CN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)