missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropurine riboside, 98%, Thermo Scientific Chemicals

Product Code. 15476479
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476479 50 g 50g
15486479 100 g 100g
15496479 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15476479 Supplier Thermo Scientific Alfa Aesar Supplier No. J64612.18
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2004-06-0
Molecular Formula C10H11ClN4O4
Molecular Weight (g/mol) 286.672
MDL Number MFCD00005738
InChI Key XHRJGHCQQPETRH-VKZRYZQFSA-N
Synonym 6-chloropurine ribonucleoside
PubChem CID 92022056
IUPAC Name (3S,4R,5S)-2-(6-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES C1=NC2=C(C(=N1)Cl)N=CN2C3C(C(C(O3)CO)O)O

Specifications

Melting Point 158°C to 162°C
Color Yellow to Green
Quantity 50 g
Beilstein 40573
Formula Weight 286.67
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Chloropurine riboside
compliance-icons
Product Identifier
  • 6-Chloropurine riboside
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 6-chloro-9-ribofuranosyl-9H-purine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.