accessibility menu, dialog, popup

UserName

CAS RN 2009-64-5

H2NNONNNH(S)OH(S)HOOH

CAS RN 2009-64-5

IUPAC Name: 2-amino-6-[(1S,2S)-1,2,3-trihydroxypropyl]-1,4-dihydropteridin-4-one
Synonyms: neopterin
Molecular Weight (g/mol): 253.22
Molecular Formula: C9H11N5O4
InChi Key: BMQYVXCPAOLZOK-NJGYIYPDSA-N
SMILES: NC1=NC(=O)C2=NC(=CN=C2N1)[C@H](O)[C@@H](O)CO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results