missing translation for 'onlineSavingsMsg'
Learn More

Neopterin, MedChemExpress

Product Code. 16400489
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16400489

Brand: Medchem Express HYW040055

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Neopterin (D-(+)-Neopterin), a catabolic product of guanosine triphosphate (GTM), serves as a marker of cellular immune system activation.

Neopterin (D-(+)-Neopterin), a catabolic product of guanosine triphosphate (GTM), serves as a marker of cellular immune system activation.

Specifications

Chemical Name or Material Neopterin
CAS 2009-64-5
Color White
Purity Grade Notes Research
Molecular Formula C9H11N5O4
Quantity 10 mg
Synonym D-(+)-Neopterin D-erythro-Neopterin
Solubility Information DMSO : 6.25 mg/mL (24.68 mM; Need ultrasonic)
SMILES OC1=C2C(N=CC([C@H](O)[C@H](O)CO)=N2)=NC(N)=N1
Molecular Weight (g/mol) 253.21
Formula Weight 253.21
Percent Purity 98.03%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.