accessibility menu, dialog, popup

UserName

CAS RN 20098-17-3

Cl(R)O(S)

CAS RN 20098-17-3

IUPAC Name: (1R,3S)-5-chloroadamantan-2-one
Synonyms: (1R,3S)-5-chloroadamantan-2-one
Molecular Weight (g/mol): 184.66
Molecular Formula: C10H13ClO
InChi Key: JPEOUSFBWXVGFX-VZCHMASFSA-N
SMILES: ClC12CC3C[C@@H](C1)C(=O)[C@H](C3)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results