CAS RN 2011-48-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(4-Methoxybenzyl)hydrazine Hydrochloride, TRC
CAS: 2011-48-5 Molecular Formula: C8H12N2O . HCl Molecular Weight (g/mol): 188.65 Synonym: 1-(4-Methoxybenzyl)hydrazine Hydrochloride,[(4-Methoxyphenyl)methyl]hydrazine Hydrochloride IUPAC Name: (4-methoxyphenyl)methylhydrazine;hydrochloride SMILES: Cl.COc1ccc(CNN)cc1