missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxybenzyl)hydrazine Hydrochloride, TRC

Product Code. 30472042
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472042 5 g 5g
30471030 1 g 1g
30471318 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30472042 Supplier Toronto Research Chemicals Supplier No. TRCM3311735G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2011-48-5
Molecular Formula C8H12N2O . HCl
Molecular Weight (g/mol) 188.65
Synonym 1-(4-Methoxybenzyl)hydrazine Hydrochloride, [(4-Methoxyphenyl)methyl]hydrazine Hydrochloride
IUPAC Name (4-methoxyphenyl)methylhydrazine;hydrochloride
SMILES Cl.COc1ccc(CNN)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H12N2O.ClH/c1-11-8-4-2-7(3-5-8)6-10-9;/h2-5,10H,6,9H2,1H3;1H
Formula Weight 188.0716
Chemical Name or Material (4-Methoxybenzyl)hydrazine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.