accessibility menu, dialog, popup

UserName

H3CONOH3CHCl

CAS RN 20232-39-7

IUPAC Name: 6,7-dimethoxy-3,4-dihydroisoquinolin-2-ium chloride
Synonyms: 6,7-dimethoxy-3,4-dihydroisoquinolin-2-ium chloride
Molecular Weight (g/mol): 227.69
Molecular Formula: C11H14ClNO2
InChi Key: PQXVEYYRJHMTEV-UHFFFAOYSA-N
SMILES: [Cl-].COC1=CC2=C(C=[NH+]CC2)C=C1OC
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)