missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate, 99+%, Thermo Scientific™

Product Code. 11916501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11916501 25 g Glass Bottle 25g
10723251 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916501 Supplier Thermo Scientific Acros Supplier No. 265960250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 20232-39-7
Molecular Formula C11H14ClNO2
Molecular Weight (g/mol) 227.69
MDL Number MFCD11099370
InChI Key PQXVEYYRJHMTEV-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, chloride, 6,7-dimethoxy-3,4-dihydro-isoquinoline hydrochloride, pubchem17358, ksc496s5h, 3,4-dihydro-6,7-dimethoxyisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hydro-chloride, 6,7-dimethoxy-3,4-dihydroisoquinolinehydrochloridehydrate
PubChem CID 2724664
SMILES Cl.COC1=CC2=C(C=NCC2)C=C1OC

Specifications

Melting Point 198.0°C to 202.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (on dry substance) (Argentometry)
Packaging Glass Bottle
Quantity 25 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline;hydrochloride
Formula Weight 227.69
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 6, 7-Dimethoxy-3, 4-dihydroisoquinoline hydrochloride hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.