accessibility menu, dialog, popup

UserName

CAS RN 2024-83-1

H3COOCH3N

CAS RN 2024-83-1

IUPAC Name: 3,4-dimethoxybenzonitrile
Synonyms: 3,4-dimethoxybenzonitrile
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: OSEQIDSFSBWXRE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results