accessibility menu, dialog, popup

UserName

CAS RN 202752-04-3

IUPAC Name: 4'-methoxy-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: 4'-methoxy-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 282.74
Molecular Formula: C13H11ClO3S
InChi Key: AJWXJCPSCXFPJJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)