missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxy(1,1'-biphenyl)-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10219502
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10219502 1 g Amber Glass Bottle 1g
10094972 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10219502 Supplier Thermo Scientific Maybridge Supplier No. MO07016DA
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 202752-04-3
Molecular Formula C13H11ClO3S
Molecular Weight (g/mol) 282.74
MDL Number MFCD01631917
InChI Key AJWXJCPSCXFPJJ-UHFFFAOYSA-N
Synonym 4'-methoxybiphenyl-4-sulfonyl chloride, 4'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonyl chloride, 4'-methoxy-biphenyl-4-sulfonyl chloride, 4'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl phenyl sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-methoxy, 4'-methoxy biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonylchloride, 4-4-methoxyphenyl phenyl sulfonylchloride
PubChem CID 2794750
IUPAC Name 4-(4-methoxyphenyl)benzenesulfonyl chloride
SMILES COC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 282.75
Percent Purity 97%
Chemical Name or Material 4';-Methoxy[1, 1';-biphenyl]-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.