accessibility menu, dialog, popup

UserName

CAS RN 202865-71-2

OOBrF(E)

CAS RN 202865-71-2

IUPAC Name: (2E)-3-(5-bromo-2-fluorophenyl)prop-2-enoate
Synonyms: (2E)-3-(5-bromo-2-fluorophenyl)prop-2-enoate
Molecular Weight (g/mol): 244.04
Molecular Formula: C9H5BrFO2
InChi Key: ROTBALNVXDGIQY-DAFODLJHSA-M
SMILES: [O-]C(=O)\C=C\C1=CC(Br)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results