accessibility menu, dialog, popup

UserName

CAS RN 202982-72-7

OHICl

CAS RN 202982-72-7

IUPAC Name: 4-chloro-3-iodophenol
Synonyms: 4-chloro-3-iodophenol
Molecular Weight (g/mol): 254.45
Molecular Formula: C6H4ClIO
InChi Key: UPIATGHEVZKVRT-UHFFFAOYSA-N
SMILES: OC1=CC(I)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results