CAS RN 203645-59-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Fenitrothion-d6, TRC
CAS: 203645-59-4 Molecular Formula: C9 2H6 H6 N O5 P S Molecular Weight (g/mol): 283.27 Synonym: Phosphorothioic acid, O,O-di(methyl-d3) O-(3-methyl-4-nitrophenyl) ester (9CI),Fenitrothion D6 (O,O-dimethyl D6) IUPAC Name: (3-methyl-4-nitrophenoxy)-sulfanylidene-bis(trideuteriomethoxy)-λ5-phosphane SMILES: [2H]C([2H])([2H])OP(=S)(Oc1ccc(c(C)c1)[N+](=O)[O-])OC([2H])([2H])[2H]
2
Fenitrothion-d6 (O,O-dimethyl-d6), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.