accessibility menu, dialog, popup

UserName

ClCH2

CAS RN 2039-85-2

IUPAC Name: 1-chloro-3-ethenylbenzene
Synonyms: M-chlorostyrene
Molecular Weight (g/mol): 138.59
Molecular Formula: C8H7Cl
InChi Key: BOVQCIDBZXNFEJ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(C=C)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results