accessibility menu, dialog, popup

UserName

BrH2C

CAS RN 2039-88-5

IUPAC Name: 1-bromo-2-ethenylbenzene
Synonyms: 2-bromostyrene
Molecular Weight (g/mol): 183.05
Molecular Formula: C8H7Br
InChi Key: SSZOCHFYWWVSAI-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results