accessibility menu, dialog, popup

UserName

CAS RN 2042-14-0

IUPAC Name: 4-methyl-3-nitrophenol
Synonyms: 3-nitro-P-cresol
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: BQEXDUKMTVYBRK-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results