accessibility menu, dialog, popup

UserName

BrN

CAS RN 2042-37-7

IUPAC Name: 2-bromobenzonitrile
Synonyms: 2-bromobenzonitrile
Molecular Weight (g/mol): 182.02
Molecular Formula: C7H4BrN
InChi Key: AFMPMSCZPVNPEM-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)