accessibility menu, dialog, popup

UserName

CAS RN 20443-74-7

IUPAC Name: 4'-chloro-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: 4'-chloro-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 287.15
Molecular Formula: C12H8Cl2O2S
InChi Key: NWYUSJMIHFIMTA-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results