accessibility menu, dialog, popup

UserName

H3COOONH3C

CAS RN 20485-39-6

IUPAC Name: ethyl 4-methyl-1,3-oxazole-5-carboxylate
Synonyms: ethyl 4-methyl-1,3-oxazole-5-carboxylate
Molecular Weight (g/mol): 155.15
Molecular Formula: C7H9NO3
InChi Key: XNMORZSEENWFLI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=CO1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results