accessibility menu, dialog, popup

UserName

CH3SnH3CCH3NN

CAS RN 205371-27-3

IUPAC Name: 2-(tributylstannyl)pyrazine
Synonyms: 2-(tributylstannyl)pyrazine
Molecular Weight (g/mol): 369.14
Molecular Formula: C16H30N2Sn
InChi Key: OVBXTKIWZAHFAC-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC=CN=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results