missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)pyrazine, 95%

Product Code. 15498077
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498077 1 g 1g
15408087 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498077 Supplier Thermo Scientific Alfa Aesar Supplier No. H51370.03
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 205371-27-3 | C16H30N2Sn | 369.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205371-27-3
Molecular Formula C16H30N2Sn
Molecular Weight (g/mol) 369.14
MDL Number MFCD01319077
InChI Key OVBXTKIWZAHFAC-UHFFFAOYSA-N
Synonym 2-tributylstannyl pyrazine, 2-pyrazinyl tributyltin, 2-tributylstannylpyrazine, pyrazine, tributylstannyl, pyrazine,2-tributylstannyl, pyrazin-2-yl tributylstannane, tributylstannyl-pyrazine, 2-tri-n-butylstannyl pyrazine, tributyl-pyrazin-2-yl-stannane, tributylstannylpyrazine
PubChem CID 2737316
IUPAC Name tributyl(pyrazin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=CN=C1

Specifications

Density 1.171
Boiling Point 130°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.516
Quantity 1 g
UN Number UN2788
Formula Weight 369.12
Percent Purity 95%
Chemical Name or Material 2-(Tri-n-butylstannyl)pyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.