CAS RN 2054858-98-7
CAS RN 2054858-98-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5,7-Dichloro Hydroxychloroquine, TRC
CAS: 2054858-98-7 Molecular Formula: C18H25Cl2N3O Molecular Weight (g/mol): 370.32 Synonym: 2-((4-((5,7-Dichloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol SMILES: CCN(CCO)CCCC(NC1=C2C(Cl)=CC(Cl)=CC2=NC=C1)C