missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloro Hydroxychloroquine, TRC

Product Code. 30378822
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378822 100 mg
30371377 50 mg
30378070 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378822 Supplier Toronto Research Chemicals Supplier No. TRCD432520100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2054858-98-7
Molecular Formula C18H25Cl2N3O
Molecular Weight (g/mol) 370.32
Synonym 2-((4-((5,7-Dichloroquinolin-4-yl)amino)pentyl)(ethyl)amino)ethanol
SMILES CCN(CCO)CCCC(NC1=C2C(Cl)=CC(Cl)=CC2=NC=C1)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H25Cl2N3O/c1-3-23(9-10-24)8-4-5-13(2)22-16-6-7-21-17-12-14(19)11-15(20)18(16)17/h6-7,11-13,24H,3-5,8-10H2,1-2H3,(H,21,22)
Chemical Name or Material 5,7-Dichloro Hydroxychloroquine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.